MilliporeSigma
  • Three-dimensional structure of the ligand-binding core of GluR2 in complex with the agonist (S)-ATPA: implications for receptor subunit selectivity.
Three-dimensional structure of the ligand-binding core of GluR2 in complex with the agonist (S)-ATPA: implications for receptor subunit selectivity.
Journal of medicinal chemistry (2003-02-21)
Marie-Louise Lunn, Anders Hogner, Tine B Stensbøl, Eric Gouaux, Jan Egebjerg, Jette S Kastrup
RÉSUMÉ
Two X-ray structures of the GluR2 ligand-binding core in complex with (S)-2-amino-3-(5-tert-butyl-3-hydroxy-4-isoxazolyl)propionic acid ((S)-ATPA) have been determined with and without Zn(2+) ions. (S)-ATPA induces a domain closure of ca. 21 degrees compared to the apo form. The tert-butyl moiety of (S)-ATPA is buried in a partially hydrophobic pocket and forces the ligand into the glutamate-like binding mode. The structures provide new insight into the molecular basis of agonist selectivity between AMPA and kainate receptors.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
ATPA, solid