MilliporeSigma
  • Kalopanaxsaponin A from Kalopanax pictus, a potent antioxidant in the rheumatoidal rat treated with Freund's complete adjuvant reagent.
Kalopanaxsaponin A from Kalopanax pictus, a potent antioxidant in the rheumatoidal rat treated with Freund's complete adjuvant reagent.
Journal of ethnopharmacology (2001-12-18)
Jongwon Choi, Keun Huh, Suk Hwan Kim, Kyung Tae Lee, Hyeong Kyu Lee, Hee Juhn Park
RÉSUMÉ
The stem bark of Kalopanax pictus is an anti-rheumatoidal arthritis drug in Oriental medicine. In the rheumatoidal rat, induced by Freund's complete adjuvant (FCA) reagent, we investigated the effects of hederagenin monodesmosides of K. pictus on oxidative stress and hepatic drug-metabolizing enzymes. Kalopanaxsaponin-A (KPS-A) significantly decreased malondialdehyde formation and the activities of xanthine oxidase and aldehyde oxidase of hepatic non-microsomal systems in FCA reagent-treated rats. In addition, increased activity levels of superoxide dismutase, catalase and glutathione peroxidase were also observed. The effects of KPS-A were more potent than the effects of KPS-I. These results suggested that KPS-A, extracted from K. pictus, could reduce rheumatoidal syndromes through antioxidative mechanisms.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Hederacoside C, analytical standard
Hederacoside C, primary reference standard