MilliporeSigma
NPS 1506, a novel NMDA receptor antagonist and neuroprotectant. Review of preclinical and clinical studies.
Annals of the New York Academy of Sciences (2000-02-11)
A L Mueller, L D Artman, M F Balandrin, E Brady, Y Chien, E G Delmar, K George, A Kierstead, T B Marriott, S T Moe, M K Newman, J L Raszkiewicz, E L Sanguinetti, B C van Wagenen, D Wells
RÉSUMÉ
NPS 1506 is a moderate affinity, uncompetitive N-methyl-D-aspartate (NMDA) receptor antagonist. NPS 1506 is neuroprotective in rodent models of ischemic stroke, hemorrhagic stroke, and head trauma, with a 2-hr window of opportunity. Neuroprotectant doses of NPS 1506 ranged from approximately 0.1-1.0 mg/kg, with peak plasma concentrations ranging from 8-80 ng/mL. Even at doses producing behavioral toxicity, NPS 1506 did not elicit MK-801-like behaviors, did not generalize to phencyclidine (PCP), and did not elicit neuronal vacuolization. In a Phase I study, intravenous (i.v.) doses of NPS 1506 from 5-100 mg were well tolerated and provided plasma concentrations in excess of those required for neuroprotection in rodents. Adverse events at the 100-mg dose included mild dizziness and lightheadedness, and mild to moderate ataxia. Neither PCP-like psychotomimetic effects nor cardiovascular effects were noted. The long plasma half-life of NPS 1506 (approximately 60 hr) suggests that a single i.v. dose will provide prolonged neuroprotection in humans.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3,3-Diphenylpropylamine, 97%