MilliporeSigma
  • N,N'-Diacetyl-L-cystine-the disulfide dimer of N-acetylcysteine-is a potent modulator of contact sensitivity/delayed type hypersensitivity reactions in rodents.
N,N'-Diacetyl-L-cystine-the disulfide dimer of N-acetylcysteine-is a potent modulator of contact sensitivity/delayed type hypersensitivity reactions in rodents.
The Journal of pharmacology and experimental therapeutics (1999-02-23)
B Särnstrand, A H Jansson, G Matuseviciene, A Scheynius, S Pierrou, H Bergstrand
RÉSUMÉ
Oral N-acetyl-L-cysteine (NAC) is used clinically for treatment of chronic obstructive pulmonary disease. NAC is easily oxidized to its disulfide. We show here that N,N'-diacetyl-L-cystine (DiNAC) is a potent modulator of contact sensitivity (CS)/delayed type hypersensitivity (DTH) reactions in rodents. Oral treatment of BALB/c mice with 0.003 to 30 micromol/kg DiNAC leads to enhancement of a CS reaction to oxazolone; DiNAC is 100 to 1000 times more potent than NAC in this respect, indicating that it does not act as a prodrug of NAC. Structure-activity studies suggest that a stereochemically-defined disulfide element is needed for activity. The DiNAC-induced enhancement of the CS reaction is counteracted by simultaneous NAC-treatment; in contrast, the CS reaction is even more enhanced in animals treated with DiNAC together with the glutathione-depleting agent buthionine sulfoximine. These data suggest that DiNAC acts via redox processes. Immunohistochemically, ear specimens from oxazolone-sensitized and -challenged BALB/c mice treated with DiNAC display increased numbers of CD8(+) cells. DiNAC treatment augments the CS reaction also when fluorescein isothiocyanate is used as a sensitizer in BALB/c mice; this is a purported TH2 type of response. However, when dinitrofluorobenzene is used as a sensitizer, inducing a purported TH1 type of response, DiNAC treatment reduces the reaction. Treatment with DiNAC also reduces a DTH footpad-swelling reaction to methylated BSA. Collectively, these data indicate that DiNAC in vivo acts as a potent and effective immunomodulator that can either enhance or reduce the CS or DTH response depending on the experimental conditions.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Acetylcysteine impurity C, European Pharmacopoeia (EP) Reference Standard