MilliporeSigma
Prevention of canine leishmaniosis in a hyper-endemic area using a combination of 10% imidacloprid/4.5% flumethrin.
PloS one (2013-03-02)
Domenico Otranto, Filipe Dantas-Torres, Donato de Caprariis, Giancarlo Di Paola, Viviana D Tarallo, Maria S Latrofa, Riccardo P Lia, Giada Annoscia, Edward B Breitshwerdt, Cinzia Cantacessi, Gioia Capelli, Dorothee Stanneck
RÉSUMÉ
Dogs are the main reservoir hosts of Leishmania infantum, the agent of human zoonotic visceral leishmaniosis. This study investigated the efficacy of a polymer matrix collar containing a combination of 10% imidacloprid and 4.5% flumethrin as a novel prophylactic measure to prevent L. infantum infections in young dogs from a hyper-endemic area of southern Italy, with a view towards enhancing current control strategies against both human and canine leishmaniosis. The study was carried out on 124 young dogs, of which 63 were collared (Group A) while 61 were left untreated (Group B), from March-April 2011 until March 2012. Blood and skin samples were collected at baseline (April 2011) and at the first, second, third and fourth follow-up time points (July, September 2011 and November 2011, and March 2012, respectively). Bone marrow and conjunctiva were sampled at baseline and at the fourth follow-up. Serological, cytological and molecular tests were performed to detect the presence of L. infantum in the different tissues collected. At the end of the trial, no dog from Group A proved positive for L. infantum at any follow-up, whereas 22 dogs from Group B were infected (incidence density rate = 45.1%); therefore, the combination of 10% imidacloprid and 4.5% flumethrin was 100% efficacious for the prevention of L. infantum infection in young dogs prior to their first exposure to the parasite in a hyper-endemic area for CanL. The use of collars containing 10% imidacloprid and 4.5% flumethrin conferred long-term protection against infection by L. infantum to dogs located in a hyper-endemic area, thus representing a reliable and sustainable strategy to decrease the frequency and spread of this disease among the canine population which will ultimately result in the reduction of associated risks to human health.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
Flumethrin, PESTANAL®, analytical standard