MilliporeSigma
  • Synthesis of hyaluronic-acid-related oligosaccharides and analogues, as their 4-methoxyphenyl glycosides, having N-acetyl-beta-D-glucosamine at the reducing end.
Synthesis of hyaluronic-acid-related oligosaccharides and analogues, as their 4-methoxyphenyl glycosides, having N-acetyl-beta-D-glucosamine at the reducing end.
Carbohydrate research (1998-09-19)
K M Halkes, T M Slaghek, T K Hyppönen, P H Kruiskamp, T Ogawa, J P Kamerling, J F Vliegenthart
RÉSUMÉ
To contribute to the possibilities to study the ability of oligosaccharide fragments of hyaluronic acid to induce angiogenesis, several hyaluronic-acid-related oligosaccharides and their 6-O-sulfated analogues were synthesised as their 4-methoxyphenyl glycosides having 2-acetamido-2-deoxy-D-glucopyranose at the reducing end. In all syntheses described, the D-glucopyranosyluronic acid residue was obtained by oxidation at C-6 of a corresponding D-glucopyranosyl residue after construction of the oligosaccharide backbone, using pyridinium dichromate and acetic anhydride.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Pyridinium dichromate, 98%