MilliporeSigma
Peptide mimics as substrates for the intestinal peptide transporter.
The Journal of biological chemistry (1998-02-07)
C S Temple, A K Stewart, D Meredith, N A Lister, K M Morgan, I D Collier, R D Vaughan-Jones, C A Boyd, P D Bailey, J R Bronk
RÉSUMÉ
4-Aminophenylacetic acid (4-APAA), a peptide mimic lacking a peptide bond, has been shown to interact with a proton-coupled oligopeptide transporter using a number of different experimental approaches. In addition to inhibiting transport of labeled peptides, these studies show that 4-APAA is itself translocated. 4-APAA transport across the rat intact intestine was stimulated 18-fold by luminal acidification (to pH 6.8) as determined by high performance liquid chromatography (HPLC); in enterocytes isolated from mouse small intestine the intracellular pH was reduced on application of 4-APAA, as shown fluorimetrically with the pH indicator carboxy-SNARF; 4-APAA trans-stimulated radiolabeled peptide transport in brush-border membrane vesicles isolated from rat renal cortex; and in Xenopus oocytes expressing PepT1, 4-APAA produced trans-stimulation of radiolabeled peptide efflux, and as determined by HPLC, was a substrate for translocation by this transporter. These results with 4-APAA show for the first time that the presence of a peptide bond is not a requirement for rapid translocation through the proton-linked oligopeptide transporter (PepT1). Further investigation will be needed to determine the minimal structural requirements for a molecule to be a substrate for this transporter.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Aminophenylacetic acid, 98%