MilliporeSigma
  • Non-phenolic linear diarylheptanoids from Curcuma xanthorrhiza: a novel type of topical anti-inflammatory agents: structure-activity relationship.
Non-phenolic linear diarylheptanoids from Curcuma xanthorrhiza: a novel type of topical anti-inflammatory agents: structure-activity relationship.
Planta medica (1996-06-01)
P Claeson, U Pongprayoon, T Sematong, P Tuchinada, V Reutrakul, P Soontornsaratune, W C Taylor
RÉSUMÉ
The topical anti-inflammatory activity of three non-phenolic linear 1,7-diarylheptanoids, previously isolated from a Thai medicinal plant, Curcuma xanthorrhiza (Zingiberaceae) and four new semi-synthetic derivatives of the naturally occurring compounds were assessed in the murine model of ethyl phenylpropiolate-induced ear edema. The naturally occurring compound 1E,3E,1,7-diphenylheptadien-5-one (6) exerted the most potent anti-inflammatory activity, with an ID50 value of similar magnitude to that of the reference drug oxyphenbutazone (67 vs. 46 micrograms/ear, respectively). None of the semi-synthetic diarylheptanoids was more active than 6. The chemical structures and pharmacological data of the natural and semi-synthetic derivatives identified a distinct structure-activity relationship. The degree of unsaturation in positions 1 and 3, and the nature of the oxygenated functional group in position 5 of the C7-chain were found to play significant roles in determining the observed in vivo activity. Based on these findings, the non-phenolic linear 1,7-diarylheptanoids-are proposed to represent a novel class of topical anti-inflammatory agents.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ethyl phenylpropiolate, 98%