MilliporeSigma
Studies of the tumorigenic potential of 4-substituted phenylhydrazines by the subcutaneous route.
Journal of toxicology and environmental health (1981-07-01)
B Toth, D Nagel
RÉSUMÉ
4-Methylphenylhydrazine hydrochloride (4-MPH) was administered to randomly bred Swiss mice as 26 weekly sc injections of 140 microgram per gram of body weight and N'-acetyl-4-(hydroxymethyl)phenylhydrazine (AMPH) as 26 weekly sc injections of 500 microgram/g. As a solvent control, physiological saline was also given as 26 weekly sc injections of 0.01 ml/g. The 4-MPH treatment induced a significant incidence (24%) of fibrosarcomas in males. In the 4-MPH-treated females and some AMPH-treated male mice, a few soft-tissue tumors were observed; however, their appearance could not be related to treatment. 4-MPH is formed under special experimental conditions from 4-hydroxymethylphenyl-hydrazine, which is an in vitro breakdown product of agaritine, an ingredient of the cultivated mushroom Agaricus bisporus. The implications of the findings are discussed.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
p-Tolylhydrazine hydrochloride, 98%