MilliporeSigma
Silicon compounds as substrates and inhibitors of acetylcholinesterase.
Biochimica et biophysica acta (1984-12-07)
A Aberman, D Segal, Y Shalitin, A L Gutman
RÉSUMÉ
Several trimethylsilyl derivatives were found to be ligands of acetylcholinesterase (acetylcholine acetylhydrolase, EC 3.1.1.7): trimethylsilylethyl acetate (III) and trimethylsilylmethyl acetate (V) are substrates of the enzyme, whereas trimethylsilylethanol (VIII) is a competitive inhibitor. The silicon compounds have kinetic parameters similar to those of their carbon analogues, except for trimethylsilylmethyl acetate, which is a substrate of acetylcholinesterase, whereas its carbon analogue is not susceptible to enzymic hydrolysis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-(Trimethylsilyl)ethanol, 96%