MilliporeSigma
  • Aromatic hydrocarbon metabolites in fish: automated extraction and high-performance liquid chromatographic separation into conjugate and non-conjugate fractions.
Aromatic hydrocarbon metabolites in fish: automated extraction and high-performance liquid chromatographic separation into conjugate and non-conjugate fractions.
Journal of chromatography (1982-02-19)
M M Krahn, T K Collier, D C Malins
RÉSUMÉ
An automated extractor-concentrator was used to extract metabolites of naphthalene, 2,6-dimethylnaphthalene, and benzo[a]pyrene from serum, bile and liver homogenate of rainbow trout (Salmo gairdneri). The extracts were analyzed by reversed-phase high-performance liquid chromatography (HPLC) with fluorescence detection. Recoveries of naphthalene and 2,6-dimethylnaphthalene metabolites from all matrices were generally greater than 90%; however, the recoveries of benzo[a]pyrene metabolites from serum ranged from 37-99%. In addition, conjugated metabolites of polycyclic aromatic hydrocarbons (PAHs) were separated from non-conjugated metabolites and parent PAHs by using two diol columns with normal-phase HPLC. The extraction and separation techniques were also applied to isolate metabolites in samples from fish fed 2,6-dimethylnaphthalene.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,6-Dimethylnaphthalene, 99%