MilliporeSigma
Inhibition of tolbutamide metabolism by substituted imidazole drugs in vivo: evidence for a structure-activity relationship.
British journal of pharmacology (1985-05-01)
D J Back, J F Tjia
RÉSUMÉ
Tolbutamide has been used as a model drug for an examination of the effects of eleven substituted imidazole compounds on hepatic metabolism in vivo. The 1-substituted compounds 1-methylimidazole, miconazole, clotrimazole and ketoconazole produced marked alterations in tolbutamide kinetics (increased half-life, decreased clearance). However, if there was substitution in the 2- position, irrespective of a substituent on N-1, then the compound did not appear to inhibit metabolism (e.g. 2-methylimidazole, 1,2-dimethylimidazole, methimazole, metronidazole). The 4- substituted compounds, 4-methylimidazole and cimetidine were inhibitors. A structure-activity relationship for the inhibitory actions of the substituted imidazoles is thus evident in vivo.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-Hydroxytolbutamide, ≥98% (HPLC)