MilliporeSigma
[Determination of aspartame--methyl ester of L-aspartyl-L-phenyl- alanine using the amino acid analyzer].
Prikladnaia biokhimiia i mikrobiologiia (1987-05-01)
E A Timokhina, L A Liublinskaia, S E Boĭtsova, V M Stepanov
RÉSUMÉ
A technique for quantitative determining of the synthetic sweetener aspartame. NH2-Asp-Phe-OMe and its hydrolysis product, NH2-Asp-Phe-OH, is proposed based on the use of an amino acid analyzer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Asp-Phe, 96%