MilliporeSigma
The regulation of steroidogenesis is different in the two types of ovine luteal cells.
Canadian journal of physiology and pharmacology (1985-03-01)
P B Hoyer, G D Niswender
RÉSUMÉ
Ovine luteal tissue contains two distinct steroidogenic cell types, small (8-20 microns) and large (greater than 20 microns), which differ based on morphological and biochemical criteria. Unstimulated small cells secrete low levels of progesterone, respond to LH or dibutyryl cAMP (dbcAMP) with enhanced secretion of progesterone, and contain most of the receptors for LH. The unstimulated large cells, conversely, secrete high levels of progesterone, have few, if any, receptors for LH, and do not respond to LH or dbcAMP with increased progesterone secretion. The lack of response to dbcAMP by large cells was investigated. Large cells incubated in the presence of cholesterol, ram serum, or 25-hydroxycholesterol did not demonstrate substrate limitation. Hormone-independent stimulation of adenylate cyclase by cholera toxin or forskolin resulted in increased adenylate cyclase activities (P less than 0.01), cAMP accumulation (P less than 0.05), and the binding of endogenous cAMP (P less than 0.05) by type I cAMP-dependent protein kinase in both small and large cells. These treatments were accompanied by enhanced secretion of progesterone (P less than 0.05) in small cells. In contrast, large cells did not respond with an increase in progesterone secretion under these conditions. These observations suggest that the high rate of secretion of progesterone in unstimulated large cells is not regulated by cAMP.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
20α-Hydroxycholesterol, analytical standard