MilliporeSigma
[Modification of DNA with dihydrazide of succinic acid for preparation of hybridization probes].
Bioorganicheskaia khimiia (1989-10-01)
M F Turchinskiĭ, E I Aĭnbinder, V D Knorre, S N Shcherbo
RÉSUMÉ
A two-step chemical method of introduction of nonradioactive labels in DNA was proposed. At first step DNA is modified by succinic dihydrazide at pH 5.0 and 95 degrees C, or at pH 4.5 and 37 degrees C in presence of sodium bisulfite. Then FITC or biotin are joined to the hydrazide groups. DNA modified in this way were shown to be effective hybridisation probes.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Succinic dihydrazide, 96%