MilliporeSigma
Exploitation of eukaryotic subcellular targeting mechanisms by bacterial effectors.
Nature reviews. Microbiology (2013-04-17)
Stuart W Hicks, Jorge E Galán
RÉSUMÉ
Several bacterial species have evolved specialized secretion systems to deliver bacterial effector proteins into eukaryotic cells. These effectors have the capacity to modulate host cell pathways in order to promote bacterial survival and replication. The spatial and temporal context in which the effectors exert their biochemical activities is crucial for their function. To fully understand effector function in the context of infection, we need to understand the mechanisms that lead to the precise subcellular localization of effectors following their delivery into host cells. Recent studies have shown that bacterial effectors exploit host cell machinery to accurately target their biochemical activities within the host cell.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Myristoleic acid, ≥99% (capillary GC)