MilliporeSigma
3-Hydroxyphthalic anhydride-modified human serum albumin as a microbicide candidate inhibits HIV infection by blocking viral entry.
The Journal of antimicrobial chemotherapy (2012-12-12)
Lin Li, Jiayin Qiu, Lu Lu, Shengli An, Pengyuan Qiao, Shibo Jiang, Shuwen Liu
RÉSUMÉ
We recently demonstrated that both 3-hydroxyphthalic anhydride (HP)- and maleic anhydride-modified chicken ovalbumin (OVA) could effectively inhibit HIV-1 infection. But because OVA may cause allergy in some human subjects, here we replaced OVA with human serum albumin (HSA) in designing a new anti-HIV-1 agent, HP-HSA, and then tested its anti-HIV-1 activity and cytotoxicity. The in vitro anti-HIV-1 activities of HP-HSA were detected by measuring p24 production and luciferase activity. The cytotoxicities of HP-HSA on target cells and human vaginal and cervical epithelial cells and the effect of HP-HSA on human peripheral blood mononuclear cell (PBMC) proliferation were evaluated by XTT assay. The effect of HP-HSA on interferon-γ secretion by PBMCs was detected by enzyme-linked immunospot (ELISPOT) assay. We found that HP-HSA exhibited broad and potent antiviral activity against infection by the HIV-1 strains tested, including drug-resistant strains. HP-HSA displayed no or low cytotoxicity on human vaginal and cervical epithelial cells and the cells used for testing HIV-1 infectivity. In addition, HP-HSA had no significant effect on proliferation or interferon-γ secretion by normal or phytohaemagglutinin-stimulated human PBMCs. A time-of-addition assay indicated that HP-HSA was an HIV-1 entry inhibitor. Because of its broad and potent anti-HIV-1 activity, low cytotoxicity and low immunogenicity to humans, HP-HSA has great potential for further development as a microbicide to prevent the sexual transmission of HIV.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
3-Hydroxyphthalic anhydride, 98%