MilliporeSigma
  • Visible and NIR photoluminescence properties of a series of novel lanthanide-organic coordination polymers based on hydroxyquinoline-carboxylate ligands.
Visible and NIR photoluminescence properties of a series of novel lanthanide-organic coordination polymers based on hydroxyquinoline-carboxylate ligands.
Inorganic chemistry (2012-12-05)
Yan-Li Gai, Ke-Cai Xiong, Lian Chen, Yang Bu, Xing-Jun Li, Fei-Long Jiang, Mao-Chun Hong
RÉSUMÉ
A series of novel two-dimensional (2D) lanthanide coordination polymers with 4-hydroxyquinoline-2-carboxylate (H(2)hqc) ligands, [Ln(Hhqc)(3)(H(2)O)](n)·3nH(2)O (Ln = Eu (1), Tb (2), Sm (3), Nd (4), and Gd (5)) and [Ln(Hhqc)(ox)(H(2)O)(2)](n) (Ln = Eu (6), Tb (7), Sm (8), Tm (9), Dy (10), Nd (11), Yb (12), and Gd (13); H(2)ox = oxalic acid), have been synthesized under hydrothermal conditions. Complexes 1-5 are isomorphous, which can be described as a two-dimensional (2D) hxl/Shubnikov network based on Ln(2)(CO(2))(4) paddle-wheel units, and the isomorphous complexes 6-13 feature a 2D decker layer architecture constructed by Ln-ox infinite chains cross-linked alternatively by bridging Hhqc(-) ligands. The room-temperature photoluminescence spectra of complexes Eu(III) (1 and 6), Tb(III) (2 and 7), and Sm(III) (3 and 8) exhibit strong characteristic emissions in the visible region, whereas Nd(III) (4 and 11) and Yb(III) (12) complexes display NIR luminescence upon irradiation at the ligand band. Moreover, the triplet state of H(2)hqc matches well with the emission level of Eu(III), Tb(III), and Sm(III) ions, which allows the preparation of new optical materials with enhanced luminescence properties.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
4-Quinolinol, 98%