MilliporeSigma
  • New excited state intramolecular proton transfer (ESIPT) dyes based on naphthalimide and observation of long-lived triplet excited states.
New excited state intramolecular proton transfer (ESIPT) dyes based on naphthalimide and observation of long-lived triplet excited states.
Chemical communications (Cambridge, England) (2012-08-24)
Jie Ma, Jianzhang Zhao, Pei Yang, Dandan Huang, Caishun Zhang, Qiuting Li
RÉSUMÉ
A new excited state intramolecular proton transfer chromophore of naphthalimide (NI) conjugated 2-(2-hydroxyphenyl) benzothiazole (HBT) was prepared which shows red shifted absorption and long-lived triplet excited states.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-(2-Hydroxyphenyl)benzothiazole, 97%