MilliporeSigma
  • Colon distension, perceived burden and side-effects of CT-colonography for screening using hyoscine butylbromide or glucagon hydrochloride as bowel relaxant.
Colon distension, perceived burden and side-effects of CT-colonography for screening using hyoscine butylbromide or glucagon hydrochloride as bowel relaxant.
European journal of radiology (2012-06-12)
Margriet C de Haan, Thierry N Boellaard, Patrick M Bossuyt, Jaap Stoker
RÉSUMÉ
Compare colonic distension and perceived burden of CT-colonography between participants receiving hyoscine butylbromide (buscopan) and glucagon hydrochloride as bowel relaxant. Data were collected within a screening trial. Participants received 20mg buscopan intravenously or 1mg of glucagon intravenously (if buscopan contra-indicated). Colon distension per segment was assessed using a 4-point scale (prone and supine). Data on perceived burden of CT-colonography were collected using a questionnaire two weeks after the examination. Outcome measures between groups were compared using propensity score matching. We used a stratified Wilcoxon-Mann-Whitney test statistic for quantitative and Cochran-Mantel-Haenszel statistics for categorical variables. 541 participants were included: 336 (62%) received buscopan and 205 received glucagon. All buscopan recipients had an adequately distended colon, compared to 96% in the glucagon group (RR 7.31, 95% CI: 1.61-33.28). More glucagon recipients scored the insufflation as rather or extremely burdensome (25% vs. 16%; overall mean score 2.7 vs. 2.4; p<0.001) and more found the entire CT-colonography rather or extremely burdensome (14% vs. 7%; 2.2 vs. 1.9; p=0.001). Most frequently reported side effects were a dry mouth in the buscopan group (15%) and nausea in the glucagon group (13%). Compared to glucagon, premedication with buscopan results in significantly more adequately distended colons and a less burdensome procedure. When buscopan can be used, it is the preferred bowel relaxant.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(−)-Scopolamine N-butyl bromide, ≥98% (TLC), powder