MilliporeSigma
  • Heterocycle-functional gramine analogues: solvent- and catalyst-free synthesis and their inhibition activities against cell proliferation.
Heterocycle-functional gramine analogues: solvent- and catalyst-free synthesis and their inhibition activities against cell proliferation.
European journal of medicinal chemistry (2012-06-01)
Shaoyong Ke, Liqiao Shi, Xiufang Cao, Qingyu Yang, Ying Liang, Ziwen Yang
RÉSUMÉ
A series of novel gramine analogues were designed and synthesized via a convenient three-component reaction, and which were evaluated for their inhibition activities against cell proliferation. Their structures were confirmed by satisfactory spectra analyses mainly including (1)H NMR, and ESI-MS analyses. The preliminary assays indicated that some of the newly synthesized compounds displayed significantly good inhibition activities against human lung cancer (NCI-H460), hepatocellular liver carcinoma (HepG2), gastric cancer (SGC-7901 and BGC-823) cell lines compared with the control 5-Fluorouracil (5-FU), which might be developed as novel lead scaffold for potential anticancer agents.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Gramine, 97.5%