MilliporeSigma
Intracellular magnesium ion selective microelectrode based on a neutral carrier.
Analytical chemistry (1989-03-15)
Z M Hu, T Bührer, M Müller, B Rusterholz, M Rouilly, W Simon
RÉSUMÉ
A magnesium ion selective microelectrode based on a synthetic neutral carrier is presented. The selectivity of Mg2+ over Na+, K+, H+, and Ca2+ is sufficient for assays of intracellular magnesium ion activities. The microelectrodes with an optimized membrane composition have a resistance of about 5 x 10(10) omega and a 90% response time of less than or equal to 3 s for a tip diameter around 1 microns. The lifetime of the microelectrode cell assembly is longer than 1 week and the emf drift after equilibration is less than or equal to 0.3 mV/h.