MilliporeSigma
Successful immune tolerance induction to enzyme replacement therapy in CRIM-negative infantile Pompe disease.
Genetics in medicine : official journal of the American College of Medical Genetics (2012-01-13)
Yoav H Messinger, Nancy J Mendelsohn, William Rhead, David Dimmock, Eli Hershkovitz, Michael Champion, Simon A Jones, Rebecca Olson, Amy White, Cara Wells, Deeksha Bali, Laura E Case, Sarah P Young, Amy S Rosenberg, Priya S Kishnani
RÉSUMÉ
Infantile Pompe disease resulting from a deficiency of lysosomal acid α-glucosidase (GAA) requires enzyme replacement therapy (ERT) with recombinant human GAA (rhGAA). Cross-reactive immunologic material negative (CRIM-negative) Pompe patients develop high-titer antibody to the rhGAA and do poorly. We describe successful tolerance induction in CRIM-negative patients. Two CRIM-negative patients with preexisting anti-GAA antibodies were treated therapeutically with rituximab, methotrexate, and gammaglobulins. Two additional CRIM-negative patients were treated prophylactically with a short course of rituximab and methotrexate, in parallel with initiating rhGAA. In both patients treated therapeutically, anti-rhGAA was eliminated after 3 and 19 months. All four patients are immune tolerant to rhGAA, off immune therapy, showing B-cell recovery while continuing to receive ERT at ages 36 and 56 months (therapeutic) and 18 and 35 months (prophylactic). All patients show clinical response to ERT, in stark contrast to the rapid deterioration of their nontolerized CRIM-negative counterparts. The combination of rituximab with methotrexate ± intravenous gammaglobulins (IVIG) is an option for tolerance induction of CRIM-negative Pompe to ERT when instituted in the naïve setting or following antibody development. It should be considered in other conditions in which antibody response to the therapeutic protein elicits robust antibody response that interferes with product efficacy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Glucose tetrasaccharide, ≥99.0% (TLC)