MilliporeSigma
  • Effect of CO₂ and 1-octen-3-ol attractants for estimating species richness and the abundance of diurnal mosquitoes in the southeastern Atlantic forest, Brazil.
Effect of CO₂ and 1-octen-3-ol attractants for estimating species richness and the abundance of diurnal mosquitoes in the southeastern Atlantic forest, Brazil.
Memorias do Instituto Oswaldo Cruz (2011-06-10)
Gabriel Z Laporta, Maria Anice M Sallum
RÉSUMÉ
Studies have shown that both carbon dioxide (CO₂) and octenol (1-octen-3-ol) are effective attractants for mosquitoes. The objective of the present study was to evaluate the attractiveness of 1-octen-3-ol and CO₂ for diurnal mosquitoes in the southeastern Atlantic forest. A Latin square experimental design was employed with four treatments: CDC-light trap (CDC-LT), CDC-LT and 1-octen-3-ol, CDC-LT and CO₂ and CDC-LT with 1-octen-3-ol and CO₂. Results demonstrated that both CDC-CO₂ and CDC-CO₂-1-octen-3-ol captured a greater number of mosquito species and specimens compared to CDC-1-octen-3-ol; CDC-LT was used as the control. Interestingly, Anopheles (Kerteszia) sp. was generally attracted to 1-octen-3-ol, whereas Aedes serratus was the most abundant species in all Latin square collections. This species was recently shown to be competent to transmit the yellow fever virus and may therefore play a role as a disease vector in rural areas of Brazil.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Oct-1-èn-3-ol, 98%
Sigma-Aldrich
Oct-1-èn-3-ol, ≥98%, FCC, FG