MilliporeSigma
Toxicological responses of Cyprinus carpio after exposure to a commercial herbicide containing imazethapyr and imazapic.
Ecotoxicology and environmental safety (2010-11-26)
Bibiana Silveira Moraes, Bárbara Clasen, Vania Lucia Loro, Alexandra Pretto, Cândida Toni, Luis Antonio de Avila, Enio Marchesan, Sérgio Luiz de Oliveira Machado, Renato Zanella, Geovane Boschmann Reimche
RÉSUMÉ
Cyprinus carpio was exposed to imazethapyr and imazapic at laboratory and at field conditions. The laboratory experiment was carried out for 7 days and at rice field for 7, 30 and 90 days. Oxidative stress parameters and antioxidant profile were studied as well as metabolic parameters. After 7 days, brain AChE activity increases in laboratory and field, but in muscle, reduction was observed only in laboratory. At the same period, brain and muscle TBARS and liver CAT increase in the laboratory. Metabolic parameters showed changes in both conditions and exposure periods. After 30 days in rice field, brain AChE activity decreases and in muscle it was enhanced. After 90 days in field, only muscle AChE activity was reduced. The disorders in oxidative stress parameters and metabolism remained, indicating mainly a protein catabolism. This study pointed out short- and long-term effects of rice herbicides at environmentally relevant concentrations on toxicological parameters in tissues of C. carpio.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Imazethapyr, PESTANAL®, analytical standard