MilliporeSigma
  • Variable sensitivity to the glucocorticoid activity of cortisol in patients with primary adrenal insufficiency: assessment with ACTH profiles.
Variable sensitivity to the glucocorticoid activity of cortisol in patients with primary adrenal insufficiency: assessment with ACTH profiles.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme (2010-10-07)
B Ekman, J Blomgren, P-O Andersson, M Carlsson, H J Arnqvist
RÉSUMÉ
Our aim was to investigate the usefulness of circulating levels of adrenocorticotropic hormone (ACTH) and also salivary cortisol to monitor cortisone substitution in patients with Addison's disease. 13 patients with primary adrenal insufficiency (8 women and 5 men, age 44 ± 11 years) received 12.5 mg cortisone acetate orally at 16:00 h and 25 mg at 07:00 h. Blood samples for cortisol and ACTH analysis were drawn every hour for 24 h, and also every half hour between 07:00 and 12:00 h. Samples for salivary cortisol were collected in parallel. Total ACTH levels showed large inter-individual variations and a diurnal rhythm with a nadir in the early evening at 19:00 (median 19 ng/l, range 2-434 ng/l) and high levels in the early morning, with a peak around 07:30 (median 844 ng/l, range 45-2,249 ng/l). Plasma cortisol concentrations showed 2 peaks distinct in time, but variable in height, 1-2 h after intake of cortisone. Plasma cortisol correlated significantly with ln(ACTH) at 17:00 h (r=-0.56), at 10:00 h (r=-0.51), and at 10.30 h (r=-0.57). When tested at different time points, ln(ACTH) at 10:00 to 12:00 h was negatively correlated with plasma cortisol between 08:30 and 12:00 h. Plasma cortisol was highly correlated to ln(salivary cortisol) most of the time points measured, but 30-60 min after intake of cortisone acetate the correlation disappeared. In conclusion, the large interindividual variation in ACTH levels most likely indicates varying sensitivity to cortisol with a need for individualized dosing schemes. Furthermore ACTH-determinations may be useful for dose titration of cortisol.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Cortisone 21-acetate, ≥99%
Cortisone acetate, European Pharmacopoeia (EP) Reference Standard