MilliporeSigma
Copper-catalyzed asymmetric addition of arylboronates to isatins: a catalytic cycle involving alkoxocopper intermediates.
Chemical communications (Cambridge, England) (2010-08-21)
Ryo Shintani, Keishi Takatsu, Tamio Hayashi
RÉSUMÉ
A copper-catalyzed addition of arylboronates to isatins has been developed to give 3-aryl-3-hydroxy-2-oxindoles under mild conditions. The catalytic cycle of this process has been examined through a series of stoichiometric reactions and an effective asymmetric variant has also been described by the use of a chiral N-heterocyclic carbene ligand.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2-Oxindole, 97%