MilliporeSigma
A ring contraction strategy toward a diastereoselective total synthesis of (+)-bakkenolide A.
The Journal of organic chemistry (2010-04-10)
Vânia M T Carneiro, Helena M C Ferraz, Tiago O Vieira, Eloisa E Ishikawa, Luiz F Silva
RÉSUMÉ
A diastereoselective route to (+)-bakkenolide A is presented from the readily available optically active Wieland-Miescher ketone. This novel synthesis of this sesquiterpene lactone features the following as key stereoselective transformations: (i) the ring contraction reaction of a octalone mediated by thallium(III) nitrate (TTN); (ii) a hydrogenation to create the cis-fused junction; and (iii) the formation of the C7 quaternary center through an enolate intermediate. Furthermore, during this work, the absolute configuration of a trinorsesquiterpene isolated from Senecio humillimus was assigned.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Thallium(I) nitrate, 99.9% trace metals basis
Sigma-Aldrich
Thallium(I) nitrate, 99.999% trace metals basis