- Excitation-wavelength-dependent photoluminescence of a pyromellitic diimide nanowire network.
Excitation-wavelength-dependent photoluminescence of a pyromellitic diimide nanowire network.
Chemical communications (Cambridge, England) (2010-03-04)
Hairong Zhang, Xiaohe Xu, Hai-Feng Ji
PMID20198252
RÉSUMÉ
Nanowires from deposition of pyromellitic diimide (PMDI) from the gas phase and their unique excitation-wavelength-dependent photoluminescence were demonstrated. The luminescence peaks of the PMDI nanowires red-shifted as the excitation wavelength increased. The relationship between the luminescence peak and the excitation wavelength is nearly linear in a broad range of excitation.
MATÉRIAUX