MilliporeSigma
One-pot organocatalytic asymmetric synthesis of 3-nitro-1,2-dihydroquinolines by a dual-activation protocol.
Chemistry, an Asian journal (2009-10-16)
Yi-Feng Wang, Wei Zhang, Shu-Ping Luo, Bai-Lin Li, Ai-Bao Xia, Ai-Guo Zhong, Dan-Qian Xu
RÉSUMÉ
Generally, amine-catalyzed enantioselective transformations rely on chiral enamine or unsaturated iminium intermediates. Herein, we report a protocol involving dual activation by an aromatic iminium and hydrogen-bonding. An enantioselective aza-Michael-Henry domino reaction of 2-aminobenzaldehydes with nitroolefins has been developed through this protocol using primary amine thiourea catalysts to provide a variety of 3-nitro-1,2-dihydroquinolines in moderate yields and with up to 90% ee. The mechanism for the catalytic enantioselective reaction was confirmed by ESI mass spectrometric detection of the reaction intermediates. The products formed are substructures found in skeletons of important biological and pharmaceutical molecules.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2-Aminobenzaldehyde, ≥98%