MilliporeSigma
Prostaglandin E2 mediates cough via the EP3 receptor: implications for future disease therapy.
American journal of respiratory and critical care medicine (2009-09-05)
Sarah A Maher, Mark A Birrell, Maria G Belvisi
RÉSUMÉ
A significant population of patients with severe asthma and chronic obstructive pulmonary disease is less responsive to beta(2)-adrenoceptor agonists and corticosteroids, and there are possible safety issues concerning long-term use of these drugs. Inhaled prostaglandin E(2) (PGE(2)) is antiinflammatory and a bronchodilator in patients with asthma, but it also causes cough. We aimed to identify the receptor involved in PGE(2)-induced sensory nerve activation and cough using a range of in vitro and in vivo techniques. Depolarization of vagal sensory nerves (human, mouse, and guinea pig) was assessed as an indicator of sensory nerve acitivity. Cough was measured in a conscious guinea pig model. Using an extensive range of pharmacological tools, we identified that the EP(3) receptor mediates PGE(2)-induced depolarization of sensory nerves in human, mouse, and guinea pig. Further supporting evidence comes from data showing that responses to PGE(2) are virtually abolished in isolated vagus nerves from EP(3)-deficient mice (Ptger3(-/-)). Finally, we demonstrated the role of the EP(3) receptor in vivo using a selective EP(3) antagonist to attenuate PGE(2)-induced cough. Identification of the receptor mediating PGE(2)-induced cough represents a key step in developing a drug that is antiinflammatory and a bronchodilator but without unwanted side effects.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid