MilliporeSigma
  • A blocked diketo form of avobenzone: photostability, photosensitizing properties and triplet quenching by a triazine-derived UVB-filter.
A blocked diketo form of avobenzone: photostability, photosensitizing properties and triplet quenching by a triazine-derived UVB-filter.
Photochemistry and photobiology (2008-08-05)
Cecilia Paris, Virginie Lhiaubet-Vallet, Oscar Jiménez, Carles Trullas, Miguel Angel Miranda
RÉSUMÉ
Novel sunscreens are required providing active protection in the UVA and UVB regions. On the other hand, there is an increasing concern about the photosafety of UV filters, as some of them are not sufficiently photostable. Avobenzone is one of the most frequently employed sunscreen ingredients, but it has been reported to partially decompose after irradiation. In the present work, photophysical and photochemical studies on a methylated avobenzone-derivative have shown that the diketo form is responsible for photodegradation. A transient absorption was observed at 380 nm after laser flash photolysis excitation at 308 nm. It was assigned to the triplet excited state of the diketo form, as inferred from quenching by oxygen and beta-carotene. This transient also interacted with key building blocks of biomolecules by triplet-triplet energy transfer (in the case of thymidine) or electron transfer processes (for 2'-deoxyguanosine, tryptophan and tyrosine). Irradiation of the avobenzone derivative in the presence of a triazine UV-B filter (E-35852) diminished the undesirable effects of the compound by an efficient quenching of the triplet excited state. Thus, sunscreen formulations including triplet quenchers could provide effective protection from the potential phototoxic and photoallergic effects derived from poor photostability of avobenzone.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)-1,3-propanedione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-(4-Methoxyphenyl)-3-(4-tert-butylphenyl)-1,3-propanedione, analytical standard