MilliporeSigma
  • Recommendation to include fragrance mix 2 and hydroxyisohexyl 3-cyclohexene carboxaldehyde (Lyral) in the European baseline patch test series.
Recommendation to include fragrance mix 2 and hydroxyisohexyl 3-cyclohexene carboxaldehyde (Lyral) in the European baseline patch test series.
Contact dermatitis (2008-02-19)
Magnus Bruze, Klaus Ejner Andersen, An Goossens
RÉSUMÉ
The currently used fragrance mix in the European baseline patch test series (baseline series) fails to detect a substantial number of clinically relevant fragrance allergies. To investigate whether it is justified to include hydroxyisohexyl 3-cyclohexene carboxaldehyde (Lyral) and fragrance mix 2 containing hydroxyisohexyl 3-cyclohexene carboxaldehyde, citral, farnesol, coumarin, citronellol, and alpha-hexyl cinnamal in the European baseline patch test series. Survey of the literature on reported frequencies of contact allergy and allergic contact dermatitis from fragrance mix 2 and hydroxyisohexyl 3-cyclohexene carboxaldehyde (Lyral) as well as reported results of experimental provocation test. Fragrance mix 2 has been demonstrated to be a useful additional marker of fragrance allergy with contact allergy rates up to 5% when included in various national baseline patch test series. Of the fragrance substances present in fragrance mix 2, hydroxyisohexyl 3-cyclohexene carboxaldehyde is the most common sensitizer. Contact allergy rates between 1.5% and 3% have been reported for hydroxyisohexyl 3-cyclohexene carboxaldehyde in petrolatum (pet.) at 5% from various European centres when tested in consecutive dermatitis patients. From 2008, pet. preparations of fragrance mix 2 at 14% w/w (5.6 mg/cm(2)) and hydroxyisohexyl 3-cyclohexene carboxaldehyde at 5% w/w (2.0 mg/cm(2)) are recommended for inclusion in the baseline series. With the Finn Chamber technique, a dose of 20 mg pet. preparation is recommended. Whenever there is a positive reaction to fragrance mix 2, additional patch testing with the 6 ingredients, 5 if there are simultaneous positive reactions to hydroxyisohexyl 3-cyclohexene carboxaldehyde and fragrance mix 2, is recommended.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde, mixture of isomers, ≥97.0% (GC)