- Fragrance material review on alpha-irone.
Fragrance material review on alpha-irone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
J Lalko, A Lapczynski, D McGinty, S P Bhatia, C S Letizia, A M Api
PMID18031907
RÉSUMÉ
A toxicologic and dermatologic review of alpha-irone when used as a fragrance ingredient is presented.