MilliporeSigma
Phosphine containing oligonucleotides for the development of metallodeoxyribozymes.
Chemical communications (Cambridge, England) (2007-04-05)
Loïc Ropartz, Nico J Meeuwenoord, Gijsbert A van der Marel, Piet W N M van Leeuwen, Alexandra M Z Slawin, Paul C J Kamer
RÉSUMÉ
Novel transition metal catalysts based on oligonucleotides can be easily obtained by functionalization of 5-iodouridine with phosphine ligands, resulting in good asymmetric induction in palladium catalyzed allylic nucleophilic substitution.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
5-Iodouridine, 95%