MilliporeSigma
Insecticidal activity of 2-tridecanone against the cowpea weevil Callosobruchus maculatus (Coleoptera: Bruchidae).
Anais da Academia Brasileira de Ciencias (2007-04-03)
Yussef F B Braga, Thalles B Grangeiro, Eder A Freire, Helano L Lopes, José N S Bezerra, Manoel Andrade-Neto, Mary Anne S Lima
RÉSUMÉ
The effect of 2-tridecanone vapor on the cowpea weevil (Callosobruchus maculatus) development was determined. Seeds of cowpea were infested with adults and exposed to different doses of 2-tridecanone isolated from Pilocarpus microphyllus Stapf ex Holm, a plant species native from northeastern Brazil. The pure monoterpene was evaluated both undiluted as well as in the dilutions 1:10, 1:100 and 1:1,000 (v/v). The following parameters of the cowpea weevil life cycle were analyzed in response to decreasing doses of 2-tridecanone: number of eggs laid, percentage of egg hatching on seeds, percentage of adult emergence, adult weight at emergence, mean developmental time and number of adults emerged. Vapor of 2-tridecanone caused a significant (P < 0.05) reduction in the number of eggs laid, in the percentage of eggs hatched and in the number of emerged adults in infested seeds. The fumigant insecticidal effect of 2-tridecanone was mainly due to its ovicidal activity.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2-tridécanone, ≥96%, FG
Sigma-Aldrich
2-tridécanone, 99%