MilliporeSigma
Screening and evaluation of fruit samples for four pesticide residues.
Journal of AOAC International (2005-07-09)
Cristina Blasco, Guillermina Font, Jordi Mañes, Yolanda Picó
RÉSUMÉ
The occurrence of 4 pesticide residues, imidacloprid, carbendazim, methiocarb, and hexythiazox, was assessed in several fruits (oranges, tangerines, watermelons, and date plums) from the Valencian Community (Spain). A total of 227 samples--56 oranges, 134 tangerines, 13 watermelons, and 24 date plums--were taken from an agricultural cooperative representative of the area studied during 2001. The pesticides were determined by liquid chromatography/mass spectrometry after sample extraction with ethyl acetate and anhydrous sodium sulfate. Recoveries of 4 pesticides at 2 fortification levels (0.02 and 0.2 mg/kg), the lower of which was the quantification limit, ranged from 60 to 108%. Concentrations of imidacloprid, carbendazim, methiocarb, and hexythiazox ranged from 0.02 to 0.75 mg/kg in 184 samples (54 oranges, 119 tangerines, 3 watermelons, and 8 date plums). Nineteen samples contained methiocarb or hexythiazox residues that exceeded the maximum residue limits. Calculation of the estimated daily intakes of the 4 pesticides studied and their comparison with the Acceptable Daily Intakes established by the Food and Agriculture Organization of the World Health Organization demonstrated the safety of fruit consumption and showed the importance of monitoring for pesticide residues.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Mercaptodimethur, PESTANAL®, analytical standard