MilliporeSigma
Aging increases amyloid beta-peptide-induced 8-iso-prostaglandin F2alpha release from rat brain.
Neurobiology of aging (2003-12-17)
Luigi Brunetti, Barbara Michelotto, Giustino Orlando, Lucia Recinella, Chiara Di Nisio, Giovanni Ciabattoni, Michele Vacca
RÉSUMÉ
In order to investigate whether amyloid beta-peptide-induced oxidative damage in the brain could be related to aging, we studied the release of 8-iso-prostaglandin (PG)F2alpha, a stable marker of cellular oxidative stress, in brain synaptosomes from Wistar rats of different ages (3, 6, 12, 18 months old), both basally and after amyloid beta-peptide (1-40) perfusion. We found that basal release of 8-iso-PGF2alpha was not significantly different among all age groups of rats. Either phospholipase A2 activation induced by calcium ionophore A23187 (10 nM) or amyloid beta-peptide (5 microM) did not modify isoprostane release, when these substances were used alone. In contrast, amyloid beta-peptide (1-5 microM) preincubation caused a dose-dependent increase of A23187-stimulated 8-iso-PGF2alpha release in each age group, which was also strikingly correlated to aging of rats. Furthermore, ferric ammonium sulfate stimulates isoprostane production to levels comparable to those induced by amyloid beta-peptide. In conclusion, although 8-iso-PGF2alpha production from rat brain synaptosomes is independent from aging in the basal state, aging renders neurons more vulnerable to amyloid beta-peptide-induced oxidative toxicity.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ammonium iron(III) sulfate dodecahydrate, ACS reagent, 99%
Sigma-Aldrich
Ammonium iron(III) sulfate dodecahydrate, ReagentPlus®, ≥99%