MilliporeSigma
  • [Investigation on the changing regularity of the relationship between retention index and temperature of the GDX series stationary phases].
[Investigation on the changing regularity of the relationship between retention index and temperature of the GDX series stationary phases].
Se pu = Chinese journal of chromatography (2003-01-25)
Y C Cui, C C Zhang, Z Wang, P J Hong
RÉSUMÉ
A systematical measure of the polarity scale of the Chinese porous polymer beads of this series of stationary phase has been carried out, including the McReynolds constant at 120 degrees C by using squalane as non-polar stationary phase and the relative constant to GDX-103 at 180 degrees C. Based on the above work, two kinds stationary phases GDX-101 and GDX-103 were selected, on which the changing regularity of the relationship between retention index and temperature of the five "probes" of stationary polarity determination, including benzene, butanol, 1-nitropropane, 2-pentanone and pyridine were investigated. At the same time, the changing regularity with temperature of the relative McReynolds constant delta I of GDX-101 to GDX-103 was studied. It is found that Ip-theta c curve and delta I-theta c curve all showed a turning point at the temperature of 180 degrees C and that there was a maximum value of delta I at the temperature of 180 degrees C. So we think that the retention mechanism of this kind of stationary phase changed at 180 degrees C.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Nitropropane, ≥98.5%