MilliporeSigma
  • Flow-injection chemiluminometric determination of some thioxanthene derivatives in pharmaceutical formulations and biological fluids using the [Ru(dipy)3(2+)]-Ce(IV) system.
Flow-injection chemiluminometric determination of some thioxanthene derivatives in pharmaceutical formulations and biological fluids using the [Ru(dipy)3(2+)]-Ce(IV) system.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2002-01-05)
F A Aly, S A al-Tamimi, A A Alwarthan
RÉSUMÉ
A flow-injection (FI) methodology using tris(2,2'-dipyridyl)ruthenium(II), [Ru(dipy)3(2+)], chemiluminescence (CL) was developed for the rapid and sensitive determination of three thioxanthene derivatives, namely zuclopenthixol hydrochloride, flupentixol hydrochloride and thiothixene. The method is based on the CL reaction of the studied thioxanthenes with [Ru(dipy)3(2+)] and Ce(IV) in a sulfuric acid medium. Under the optimum conditions, calibration graphs were obtained over the concentration ranges 0.002-6 migrograms/ml for zuclopenthixol hydrochloride, 0.5-15 micrograms/ml for flupentixol hydrochloride and 0.05-7.5 micrograms/ml for thiothixene. The limits of detection (s/n = 3) were 4.2 x 10(-9) mol/l zuclopenthixol hydrochloride, 2 x 10(-8) mol/l flupentixol hydrochloride and 4.5 x 10(-8) mol/l thiothixene. The method was successfully applied to the determination of these compounds in dosage forms and biological fluids.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Cesium sulfate, Grade I, ≥99%
Sigma-Aldrich
Cesium sulfate, 99.99% trace metals basis