MilliporeSigma
Caged DNA does not aggregate in high ionic strength solutions.
Bioconjugate chemistry (1999-07-20)
V S Trubetskoy, A Loomis, P M Slattum, J E Hagstrom, V G Budker, J A Wolff
RÉSUMÉ
The assembly of DNA into compact particles that do not aggregate in physiologic salt solution occurs naturally in chromatin and viral particles but has been challenging to duplicate using artificial constructs. Cross-linking amino-containing polycations in the presence of DNA with bisimidoester cross-linker leads to the formation of caged DNA particles that are stable in salt solutions. This first demonstration of caged DNA provides insight into how natural condensation processes avoid aggregation and a promising avenue for developing nonviral gene therapy vectors.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Luperox® 231, 1,1-Bis(tert-butylperoxy)-3,3,5-trimethylcyclohexane, 92%