MilliporeSigma
  • The combination of tephrosin with 2-deoxy-D-glucose enhances the cytotoxicity via accelerating ATP depletion and blunting autophagy in human cancer cells.
The combination of tephrosin with 2-deoxy-D-glucose enhances the cytotoxicity via accelerating ATP depletion and blunting autophagy in human cancer cells.
Cancer biology & therapy (2011-11-30)
Yunjin Choi, Jeong-Hyung Lee
RÉSUMÉ
2-Deoxy-D-glucose (2-DG), a synthetic glucose analog that acts as a glycolytic inhibitor, is currently under clinical evaluation for targeting tumor cells. Tephrosin (TSN), a plant rotenoid, is known as an anticancer agent. In this study, we describe that the addition of TSN to 2-DG enhanced the cytotoxic activity of 2-DG against various types of cancer cells by accelerating ATP depletion and blunting autophagy. TSN increased the sensitivity of cancer cells to the cytotoxic effect of 2-DG. The combination of TSN and 2-DG induced acceleration of intracellular ATP depletion and the drastic activation of AMP-activated protein kinase (AMPK), which resulted in the inactivation of the mammalian target of rapamycin (mTOR) pathway. Of particular interest, TSN suppressed 2-DG-induced autophagy, a cell survival process in response to nutrient deprivation. We also showed that TSN inhibited 2-DG-induced activation of elongation factor-2 kinase (eEF-2K), which has been known to regulate 2-DG-induced autophagy. Inhibition of eEF-2K by RNA interference blunted 2-DG-induced autophagy and increased the sensitivity of cancer cells to the cytotoxic effect of 2-DG. The addition of TSN to 2-DG, however, did not enhance the cytotoxic activity of 2-DG by knockdown of eEF-2K, suggesting that inhibition of eEF-2K by tephrsoin could be a critical role in the potentiating effect of TSN on the cytotoxicity of 2-DG. Furthermore, we showed that the blunted autophagy and enhanced cytotoxicity of 2-DG was accompanied by the augmentation of apoptosis. These results show that TSN may be valuable for augmenting the therapeutic efficacy of 2-DG.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tephrosin, ≥95% (LC/MS-ELSD)