MilliporeSigma
Angiotensin II receptor subtypes are coupled with distinct signal-transduction mechanisms in neurons and astrocytes from rat brain.
Proceedings of the National Academy of Sciences of the United States of America (1991-09-01)
C Sumners, W Tang, B Zelezna, M K Raizada
RÉSUMÉ
Both neurons and astrocytes contain specific receptors for angiotensin II (AII). We used selective ligands for the AT1 and AT2 types of AII receptors to investigate the expression of functional receptor subtypes in astrocyte cultures and neuron cultures from 1-day-old (neonatal) rat brain. In astrocyte cultures, competition of 125I-labeled AII (125I-AII) specific binding with AT1 (DuP753) or AT2 (PD123177, CGP42112A, [Phe(p-NH2)6]AII) selective receptor ligands revealed a potency series of AII greater than DuP753 much greater than CGP42112A greater than [Phe(p-NH2)6]AII greater than PD123177. These results suggest a predominance of the AT1 receptor subtype in neonatal astrocytes. Also, in astrocyte cultures, AII stimulated increases in inositolphospholipid hydrolysis that were significantly reduced by the AT1 receptor antagonist DuP753 but not altered by the AT2 receptor antagonist PD123177. In neonatal neuron cultures, competition of 125I-AII specific binding with the above ligands revealed a potency series of CGP42112A = AII greater than [Phe(p- NH2)6]AII greater than PD123177 much greater than DuP753. 125I-AII specific binding to neonate neuronal cultures was reduced 73-84% by 1 microM PD123177, and the residual 125I-AII specific binding was eliminated by DuP753. Also, in neuron cultures, AII induced decreases in basal cGMP that were completely blocked by PD123177 or CGP42112A but not by DuP753. Our results suggest that astrocyte cultures from neonatal rat brains contain predominantly AT1 receptors that are coupled to a stimulation of inositophospholipid hydrolysis. In contrast, neuron cultures from neonatal rat brain contain mostly AT2 receptors that are coupled to a reduction in basal cGMP levels, but a smaller population of AT1 receptors is also present in these neurons.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
CGP-42112A, ≥95%, synthetic, solid