MilliporeSigma
β-Secretase (BACE1) inhibitors with high in vivo efficacy suitable for clinical evaluation in Alzheimer's disease.
Journal of medicinal chemistry (2013-04-18)
Hans Hilpert, Wolfgang Guba, Thomas J Woltering, Wolfgang Wostl, Emmanuel Pinard, Harald Mauser, Alexander V Mayweg, Mark Rogers-Evans, Roland Humm, Daniela Krummenacher, Thorsten Muser, Christian Schnider, Helmut Jacobsen, Laurence Ozmen, Alessandra Bergadano, David W Banner, Remo Hochstrasser, Andreas Kuglstatter, Pascale David-Pierson, Holger Fischer, Alessandra Polara, Robert Narquizian
RÉSUMÉ
An extensive fluorine scan of 1,3-oxazines revealed the power of fluorine(s) to lower the pKa and thereby dramatically change the pharmacological profile of this class of BACE1 inhibitors. The CF3 substituted oxazine 89, a potent and highly brain penetrant BACE1 inhibitor, was able to reduce significantly CSF Aβ40 and 42 in rats at oral doses as low as 1 mg/kg. The effect was long lasting, showing a significant reduction of Aβ40 and 42 even after 24 h. In contrast to 89, compound 1b lacking the CF3 group was virtually inactive in vivo.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
β-Secretase inhibitor, ≥97% (HPLC), powder