MilliporeSigma
  • The selective dopamine D3 receptor antagonists SB-277011A and NGB 2904 and the putative partial D3 receptor agonist BP-897 attenuate methamphetamine-enhanced brain stimulation reward in rats.
The selective dopamine D3 receptor antagonists SB-277011A and NGB 2904 and the putative partial D3 receptor agonist BP-897 attenuate methamphetamine-enhanced brain stimulation reward in rats.
Psychopharmacology (2007-11-07)
Krista Spiller, Zheng-Xiong Xi, Xiao-Qing Peng, Amy H Newman, Charles R Ashby, Christian Heidbreder, József Gaál, Eliot L Gardner
RÉSUMÉ
We have previously reported that selective antagonism of brain D3 receptors by SB-277011A or NGB 2904 significantly attenuates cocaine- or nicotine-enhanced brain stimulation reward (BSR). In the present study, we investigated whether the selective D3 receptor antagonists SB-277011A and NGB 2904 and the putative partial D3 agonist BP-897 similarly reduce methamphetamine (METH)-enhanced BSR. Rats were trained to respond for rewarding electrical self-stimulation of the medial forebrain bundle. To assess the degree of drug-induced changes in BSR, a rate-frequency curve shift paradigm was used to measure brain-reward threshold (theta 0). METH (0.1-0.65 mg/kg, i.p.) dose-dependently lowered ( approximately 10-50%) BSR thresholds, producing an enhancement of BSR. Pretreatment with SB-277011A (12 mg/kg, but not 24 mg/kg, i.p.) significantly attenuated METH-enhanced BSR. NGB 2904 (0.1-1.0 mg/kg, but not 10 mg/kg) also attenuated METH-enhanced BSR. SB-277011A or NGB 2904 alone, at the doses tested, had no effect on BSR. Pretreatment with BP-897 (0.1-5 mg/kg) dose-dependently attenuated METH-enhanced BSR. However, when the dose was increased to 10 mg/kg, BP-897 shifted the stimulation-response curve to the right (inhibited BSR itself) in the presence or absence of METH. Selective antagonism of D3 receptors by SB-277011A or NGB 2904 attenuates METH-enhanced BSR in rats, while the METH-enhanced BSR attenuation produced by BP-897 may involve both D3 and non-D3 receptors. These findings support a potential use of selective D3 receptor antagonists for the treatment of METH addiction.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
SB-277011-A hydrochloride hydrate, ≥98% (HPLC)