MilliporeSigma
  • Mutation in exon 1f of PLEC, leading to disruption of plectin isoform 1f, causes autosomal-recessive limb-girdle muscular dystrophy.
Mutation in exon 1f of PLEC, leading to disruption of plectin isoform 1f, causes autosomal-recessive limb-girdle muscular dystrophy.
American journal of human genetics (2010-11-27)
Hulya Gundesli, Beril Talim, Petek Korkusuz, Burcu Balci-Hayta, Sebahattin Cirak, Nurten A Akarsu, Haluk Topaloglu, Pervin Dincer
RÉSUMÉ
Limb-girdle muscular dystrophy (LGMD) is a genetically heterogeneous group of inherited muscular disorders manifesting symmetric, proximal, and slowly progressive muscle weakness. Using Affymetrix 250K SNP Array genotyping and homozygosity mapping, we mapped an autosomal-recessive LGMD phenotype to the telomeric portion of chromosome 8q in a consanguineous Turkish family with three affected individuals. DNA sequence analysis of PLEC identified a homozygous c.1_9del mutation containing an initiation codon in exon 1f, which is an isoform-specific sequence of plectin isoform 1f. The same homozygous mutation was also detected in two additional families during the analysis of 72 independent LGMD2-affected families. Moreover, we showed that the expression of PLEC was reduced in the patient's muscle and that there was almost no expression for plectin 1f mRNA as a result of the mutation. In addition to dystrophic changes in muscle, ultrastructural alterations, such as membrane duplications, an enlarged space between the membrane and sarcomere, and misalignment of Z-disks, were observed by transmission electron microscopy. Unlike the control skeletal muscle, no sarcolemmal staining of plectin was detected in the patient's muscle. We conclude that as a result of plectin 1f deficiency, the linkage between the sarcolemma and sarcomere is broken, which could affect the structural organization of the myofiber. Our data show that one of the isoforms of plectin plays a key role in skeletal muscle function and that disruption of the plectin 1f can cause the LGMD2 phenotype without any dermatologic component as was previously reported with mutations in constant exons of PLEC.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
JumpStart Taq ReadyMix avec SYBR® Green, for quantitative PCR, without MgCl2 in buffer