MilliporeSigma
Enzymatic basis of thiol-stimulated secretion of porphyrins by Escherichia coli.
Journal of bacteriology (1992-02-01)
G T Javor, E F Febre
RÉSUMÉ
1-Thioglycerol (TG) stimulates the synthesis of porphyrin in aerobically growing Escherichia coli. Here the levels of delta-aminolevulinate biosynthetic enzymes in untreated and TG-treated E. coli THU and PUC2 (a mutant of THU which overproduces porphyrins in the presence of thiols) cells were determined. TG treatment elevated the activity of glutamyl-tRNA reductase in both strains. The increased activity was not caused by activation of preexisting enzymes by thiols or by oxidizing agents but was dependent on new protein synthesis.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-Thioglycérol, ≥97%