- Adherence of Streptococcus pneumoniae to human bronchial epithelial cells (BEAS-2B).
Adherence of Streptococcus pneumoniae to human bronchial epithelial cells (BEAS-2B).
Infection and immunity (1998-02-07)
J E Adamou, T M Wizemann, P Barren, S Langermann
PMID9453647
RÉSUMÉ
Pneumococcal adherence to alveolar epithelial cells and nasopharyngeal epithelial cells has been well characterized. However, the interaction of Streptococcus pneumoniae with bronchial epithelial cells has not been studied. We have now shown that pneumococci bind specifically to a human bronchial epithelial cell line (BEAS-2B cells). Pneumococci adhered to BEAS-2B cells in a time- and dose-dependent manner. These results suggest that the bronchial epithelium may serve as an additional site of attachment for pneumococci and demonstrate the utility of the BEAS-2B cell line for studying mechanisms of pneumococcal infection.
MATÉRIAUX
Référence du produit
Marque
Description du produit
BEAS-2B Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.