MilliporeSigma
Solution structure of Alg13: the sugar donor subunit of a yeast N-acetylglucosamine transferase.
Structure (London, England : 1993) (2008-06-13)
Xu Wang, Thomas Weldeghiorghis, Guofeng Zhang, Barbara Imperiali, James H Prestegard
RÉSUMÉ
The solution structure of Alg13, the glycosyl donor-binding domain of an important bipartite glycosyltransferase in the yeast Saccharomyces cerevisiae, is presented. This glycosyltransferase is unusual in that it is active only in the presence of a binding partner, Alg14. Alg13 is found to adopt a unique topology among glycosyltransferases. Rather than the conventional Rossmann fold found in all GT-B enzymes, the N-terminal half of the protein is a Rossmann-like fold with a mixed parallel and antiparallel beta sheet. The Rossmann fold of the C-terminal half of Alg13 is conserved. However, although conventional GT-B enzymes usually possess three helices at the C terminus, only two helices are present in Alg13. Titration of Alg13 with both UDP-GlcNAc, the native glycosyl donor, and a paramagnetic mimic, UDP-TEMPO, shows that the interaction of Alg13 with the sugar donor is primarily through the residues in the C-terminal half of the protein.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
(3-Acrylamidopropyl)trimethylammonium chloride solution, 75 wt. % in H2O