MilliporeSigma
Inhibition of Nuclear PTEN Tyrosine Phosphorylation Enhances Glioma Radiation Sensitivity through Attenuated DNA Repair.
Cancer cell (2019-03-05)
Jianhui Ma, Jorge A Benitez, Jie Li, Shunichiro Miki, Claudio Ponte de Albuquerque, Thais Galatro, Laura Orellana, Ciro Zanca, Rachel Reed, Antonia Boyer, Tomoyuki Koga, Nissi M Varki, Tim R Fenton, Suely Kazue Nagahashi Marie, Erik Lindahl, Timothy C Gahman, Andrew K Shiau, Huilin Zhou, John DeGroot, Erik P Sulman, Webster K Cavenee, Richard D Kolodner, Clark C Chen, Frank B Furnari
RÉSUMÉ
Ionizing radiation (IR) and chemotherapy are standard-of-care treatments for glioblastoma (GBM) patients and both result in DNA damage, however, the clinical efficacy is limited due to therapeutic resistance. We identified a mechanism of such resistance mediated by phosphorylation of PTEN on tyrosine 240 (pY240-PTEN) by FGFR2. pY240-PTEN is rapidly elevated and bound to chromatin through interaction with Ki-67 in response to IR treatment and facilitates the recruitment of RAD51 to promote DNA repair. Blocking Y240 phosphorylation confers radiation sensitivity to tumors and extends survival in GBM preclinical models. Y240F-Pten knockin mice showed radiation sensitivity. These results suggest that FGFR-mediated pY240-PTEN is a key mechanism of radiation resistance and is an actionable target for improving radiotherapy efficacy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Src Protein, active, 10 µg, Active, N-Terminal His6-tagged, recombinant, full-length, human Src. For use in Kinase Assays.